missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hemin, 95%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0219882001
Specifications
Hemin | |
Black | |
C34H32FeN4O4 | |
1 g | |
GGIDWJQWCUJYRY-UHFFFAOYSA-L | |
iron(3+) 4,20-bis(2-carboxyethyl)-9,14-diethenyl-5,10,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,â¶.1â¸,¹¹.1¹³,¹â¶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide | |
131675604 | |
0.95 |
16009-13-5 | |
7.87g/cm3 at 25°C | |
MFCD00010726 | |
chlorohemin, hemin chloride, protoferriheme, ferriheme, ferriprotoporphyrin ix chloride, ferriprotoporphyrin ix, panhematin, protohemin, protohemin ix, ferriprotoporphyrin | |
[Fe+3].CC1=C2[N-]C(C=C3N=C(C=C4[N-]C(=CC5=NC(=C2)C(C)=C5C=C)C(C)=C4C=C)C(C)=C3CCC(O)=O)=C1CCC(O)=O | |
616.50 | |
652 | |
Crystalline Powder |
Chemical Identifiers
16009-13-5 | |
616.50 | |
GGIDWJQWCUJYRY-UHFFFAOYSA-L | |
131675604 | |
[Fe+3].CC1=C2[N-]C(C=C3N=C(C=C4[N-]C(=CC5=NC(=C2)C(C)=C5C=C)C(C)=C4C=C)C(C)=C3CCC(O)=O)=C1CCC(O)=O |
C34H32FeN4O4 | |
MFCD00010726 | |
chlorohemin, hemin chloride, protoferriheme, ferriheme, ferriprotoporphyrin ix chloride, ferriprotoporphyrin ix, panhematin, protohemin, protohemin ix, ferriprotoporphyrin | |
iron(3+) 4,20-bis(2-carboxyethyl)-9,14-diethenyl-5,10,15,19-tetramethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,â¶.1â¸,¹¹.1¹³,¹â¶]tetracosa-1(20),2,4,6(24),7,9,11,13(22),14,16,18-undecaene-21,23-diide |