missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Hemostats Kelly Forceps

Fisherbrand Hemostats Kelly Forceps are primarily used to control blood flow in surgical procedures. ASTM™ standard (F899-84) stainless steel.

$6.79 - $7.45

Specifications

Product Type Hemostats Kelly Forcep
Certifications/Compliance ISO 9001, cCMP
Material Stainless Steel
Includes Serrated jaws, box lock joint
Length (English) 5.5 in.
View More Specs

Products 2
Catalog Number Mfr. No. Tip Style Price Quantity  
Catalog Number Mfr. No. Tip Style Price Quantity  
13-812-14
View Documents
N/A Curved
Each for $7.45
 
08-907
View Documents
N/A Straight
Each for $6.79
 
Description

Description

Fisherbrand Hemostats Kelly Forceps are primarily used to control blood flow in surgical procedures. ASTM™ standard (F899-84) stainless steel.

  • Fisherbrand™ Hemostats Kelly Forceps are primarily used to control blood flow in large blood vessels and tubing during surgical procedures
  • Our forceps are designed to exceed perfromance expectations-This is the Fisherbrand Advantage.
  • 3-Stage Locking System – Variable blood flow control (Stage 0: complete flow to Stage 3: no flow)
  • Finger-ring handle designed for comfort, stability and ease of use
  • Multipurpose - Useful in educational, research or healthcare setting
  • Hand polished to mirror finish
  • Made from surgical stainless steel
  • Electronically passivated for corrosion resistance
  • Can be sterilized by Eto/gamma radiation
  • Rockwell hardness (HRC): 40-45
  • Autoclavable up to 135°C
  • Latex-free

Certifications

Manufactured under ISO 9001, cCMP guidelines.

Specifications

Specifications

Hemostats Kelly Forcep
Stainless Steel
5.5 in.
ISO 9001, cCMP
Serrated jaws, box lock joint
14 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'