missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™
Commonly used buffering agent
Supplier: Fisher BioReagents FLBP299100
Description
Commonly used buffering agent
HEPES is commonly used as a buffering agent.Specifications
≤0.01(0.1M solution) at 260nm, 0.004 max. (0.1M solution) at 280nm | |
1M Solution, pH 7.3 | |
Undesignated | |
7.2 to 7.5 | |
100 mL | |
DNase-, RNase- and Protease-Free | |
C8H18N2O4S | |
Protease free | |
N-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid | |
Molecular Biology |
HEPES Buffer | |
7365-45-9,7732-18-5 | |
0.95 to 1.05 M | |
Liquid | |
DNase free | |
Pass Test | |
Poly Bottle | |
15, 4689 | |
238.197 |