missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HEPES Free Acid, >99%, Molecular Biology Reagent, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219482780

 View more versions of this product

Catalog No. ICN19482780


Explore available promotions

Description

Description

Buffering agent
Specifications

Specifications

HEPES, free acid
7365-45-9
Powder
>99%
MFCD00006158
JKMHFZQWWAIEOD-UHFFFAOYSA-N
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
23831
238.3
Free Acid
White
100 g
C8H18N2O4S
N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid)
OCCN1CCN(CCS(O)(=O)=O)CC1
238.30
CHEBI:42334
Molecular Biology Reagent

Chemical Identifiers

7365-45-9
238.30
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
OCCN1CCN(CCS(O)(=O)=O)CC1
C8H18N2O4S
MFCD00006158
N-2-Hydroxyethylpiperazine-N'-2-ethanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid)
CHEBI:42334
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 230-907-9

storageNote1 : Store desiccated.

Recommended Storage : Store at Room Temperature (15° to 30°C).

SDS
Documents

Documents

Promotions

Promotions