missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals HEPES sodium salt, 99%
CAS: 75277-39-3 | C8H17N2NaO4S | 260.28 g/mol
$102.85 - $1409.85
Chemical Identifiers
| CAS | 75277-39-3 |
|---|---|
| Molecular Formula | C8H17N2NaO4S |
| Molecular Weight (g/mol) | 260.28 |
| MDL Number | MFCD00036463 |
| InChI Key | RDZTWEVXRGYCFV-UHFFFAOYSA-M |
| Synonym | 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt |
| PubChem CID | 2724248 |
| ChEBI | CHEBI:46758 |
| IUPAC Name | sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate |
| SMILES | [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC215000250
|
Thermo Scientific Chemicals
215000250 |
25 g |
Each for $102.85
|
|
|||||
|
AC215001000
|
Thermo Scientific Chemicals
215001000 |
100 g |
Each for $255.68
|
|
|||||
|
AC215000010
|
Thermo Scientific Chemicals
215000010 |
1 kg |
Each for $1,409.85
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
| 75277-39-3 | |
| 260.28 | |
| RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
| 2724248 | |
| sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate |
| C8H17N2NaO4S | |
| MFCD00036463 | |
| 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt | |
| CHEBI:46758 | |
| [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 |
Specifications
| (0.1 M aq. soln.), (0.1 M aq. soln.) at 0nm, 0.02 at 280nm, 0.04 at 260nm | |
| 75277-39-3 | |
| 9.5 to 10.5 (1% aq. soln.) | |
| 25 g | |
| Authentic | |
| MFCD00036463 | |
| 2-[4-(2-Hydroxyethyl)-1-piperazine]ethanesulfonic acid sodium salt | |
| [Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 | |
| 260.28 | |
| CHEBI:46758 | |
| ≥98.5% (on anhydrous substance) |
| (0.1M aq. soln.) | |
| White | |
| Crystalline Powder | |
| 99% | |
| C8H17N2NaO4S | |
| Glass bottle | |
| RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
| sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate | |
| 2724248 | |
| 260.27 | |
| HEPES sodium salt, 99% |
Safety and Handling
EINECSNumber : 278-169-7
TSCA : TSCA
RUO – Research Use Only