missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HEPES Sodium Salt, Biological Buffer, High Purity, 98%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor H1084500GM
Specifications
0.05 A.U. (0.1 M Solution at 260 nm) | |
1 | |
0.04 | |
Amber Glass Bottle | |
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 | |
260.28 | |
High Purity |
75277-39-3 | |
500 g | |
C8H17N2NaO4S | |
RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate | |
98% |
Chemical Identifiers
75277-39-3 | |
260.28 | |
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate |
C8H17N2NaO4S | |
RDZTWEVXRGYCFV-UHFFFAOYSA-M | |
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.