missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptafluoro-2,3,3-trichlorobutane, 98%, Thermo Scientific™

$62.32 - $62.32

Chemical Identifiers

CAS 335-44-4
Molecular Formula C4Cl3F7
Molecular Weight (g/mol) 287.383
MDL Number MFCD00018065
InChI Key ZPGMWBFCBUKITA-UHFFFAOYSA-N
Synonym heptafluoro-2,3,3-trichlorobutane, 2,2,3-trichloroheptafluorobutane, heptafluoro-2,2,3-trichlorobutane, butane, 2,2,3-trichloro-1,1,1,3,4,4,4-heptafluoro, acmc-20aks6, 2,3-trichloroheptafluorobutane, butane,2,3-trichloro-1,1,1,3,4,4,4-heptafluoro
PubChem CID 78977
IUPAC Name 2,2,3-trichloro-1,1,1,3,4,4,4-heptafluorobutane
SMILES C(C(C(F)(F)F)(Cl)Cl)(C(F)(F)F)(F)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1112814
View Documents
Thermo Scientific Chemicals
L1112814
25 g
Each for $62.32
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

335-44-4
287.383
ZPGMWBFCBUKITA-UHFFFAOYSA-N
78977
C(C(C(F)(F)F)(Cl)Cl)(C(F)(F)F)(F)Cl
C4Cl3F7
MFCD00018065
heptafluoro-2,3,3-trichlorobutane, 2,2,3-trichloroheptafluorobutane, heptafluoro-2,2,3-trichlorobutane, butane, 2,2,3-trichloro-1,1,1,3,4,4,4-heptafluoro, acmc-20aks6, 2,3-trichloroheptafluorobutane, butane,2,3-trichloro-1,1,1,3,4,4,4-heptafluoro
2,2,3-trichloro-1,1,1,3,4,4,4-heptafluorobutane

Specifications

335-44-4
1.748
C4Cl3F7
1.353
25 g
heptafluoro-2,3,3-trichlorobutane, 2,2,3-trichloroheptafluorobutane, heptafluoro-2,2,3-trichlorobutane, butane, 2,2,3-trichloro-1,1,1,3,4,4,4-heptafluoro, acmc-20aks6, 2,3-trichloroheptafluorobutane, butane,2,3-trichloro-1,1,1,3,4,4,4-heptafluoro
C(C(C(F)(F)F)(Cl)Cl)(C(F)(F)F)(F)Cl
287.383
287.39
Heptafluoro-2,3,3-trichlorobutane
4°C
97°C to 98°C
CF3CCl2CFClCF3
MFCD00018065
1777522
ZPGMWBFCBUKITA-UHFFFAOYSA-N
2,2,3-trichloro-1,1,1,3,4,4,4-heptafluorobutane
78977
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'