Learn More
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1490722
Description
Heptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHeptafluorobutyric acid is an ion pair reagent for reverse-phase HPLC. It is used in the sequencing, synthesis, and solubilizing of proteins and peptides. It is used as mobile phase modifier for enhancement of selectivity in the HPLC analysis of histone proteins. It is an effective additive for zinc electrodeposition.
Solubility
Miscible with water, toluene, ether, ethanol and benzene. Â Â Â Â
Notes
Hygroscopic. Light sensitive. Incompatible withbases, oxidizing agents and reducing agents.. Â
Specifications
Heptafluorobutyric acid | |
-18°C | |
120°C to 121°C | |
Unpleasant | |
1.3 | |
MFCD00004171 | |
UN3265 | |
Light sensitive | |
Miscible with water,toluene,ether,ethanol and benzene. | |
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O | |
214.039 | |
CHEBI:39426 | |
99% |
375-22-4 | |
1.648 | |
None | |
C4HF7O2 | |
CF3CF2CF2CO2H | |
100 g | |
1426882 | |
heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech | |
YPJUNDFVDDCYIH-UHFFFAOYSA-N | |
2,2,3,3,4,4,4-heptafluorobutanoic acid | |
9777 | |
214.04 |
Chemical Identifiers
375-22-4 | |
214.039 | |
YPJUNDFVDDCYIH-UHFFFAOYSA-N | |
9777 | |
2,2,3,3,4,4,4-heptafluorobutanoic acid |
C4HF7O2 | |
MFCD00004171 | |
heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech | |
CHEBI:39426 | |
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: I
EINECSNumber : 206-786-3
RTECSNumber : ET4025000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only