Learn More
Heptafluorobutyric acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 172805000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Heptafluorobutyric acid | |
-17.5°C | |
1.6400g/mL | |
Authentic | |
Glass bottle | |
1.3 | |
MFCD00004171 | |
1.64 | |
Solubility in water: miscible. Other solubilities: soluble in benzene and carbon tetrachloride, miscible with acetone, ether and petroleum ether, insoluble in carbon disulfide and mineral oil | |
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O | |
214.04 | |
CHEBI:39426 | |
99% |
375-22-4 | |
Colorless to Yellow | |
120.0°C (755.0 mmHg) | |
99% | |
C4HF7O2 | |
CF3CF2CF2CO2H | |
500 g | |
heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech | |
YPJUNDFVDDCYIH-UHFFFAOYSA-N | |
2,2,3,3,4,4,4-heptafluorobutanoic acid | |
9777 | |
214.04 | |
Liquid |
Chemical Identifiers
375-22-4 | |
214.04 | |
YPJUNDFVDDCYIH-UHFFFAOYSA-N | |
9777 | |
2,2,3,3,4,4,4-heptafluorobutanoic acid |
C4HF7O2 | |
MFCD00004171 | |
heptafluorobutyric acid, perfluorobutyric acid, heptafluorobutanoic acid, perfluorobutanoic acid, butanoic acid, heptafluoro, heptafluoro-1-butanoic acid, perfluoropropanecarboxylic acid, butyric acid, heptafluoro, kyselina heptafluormaselna, kyselina heptafluormaselna czech | |
CHEBI:39426 | |
C(=O)(C(C(C(F)(F)F)(F)F)(F)F)O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 206-786-3
RUO – Research Use Only