missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptane (HPLC), Fisher Chemical™

$436.08 - $975.92

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
H350-1
SDS View Documents
Thermo Fisher Scientific
H3501
1 L Glass Bottle
Each for $436.08
Case of 6 Each for $2,616.48
 
H350-4 Edge
SDS Accessories View Documents
Thermo Fisher Scientific
H3504
4 L Glass Bottle
Each for $926.42
Case of 4 Each for $3,705.68
Edge
 
H350SK-4
SDS View Documents
Thermo Fisher Scientific
FLH350SK4
4 L Safe-Cote™ Amber Glass
Each for $975.92
Case of 4 Each for $3,903.68
 
Description

Description

Specifications

Chemical Identifiers

142-82-5
100.21
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
C7H16
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC

Specifications

-91°C
0.014 AU max. at 254nm, 0.2 AU max. at 215nm, 0.75 AU max. at 200nm
0.683g/cm³
1 L
Pass Test
Liquid
5ppm max.
C7H16
MFCD00009544
0.4 mPa/s at 20°C
0.02% max.
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.2
Mobile phase for HPLC applications
142-82-5,28729-52-4,26635-64-3,108-87-2
Colorless
98°C
−4°C (25°F)
Glass Bottle
1.3845 to 1.3865 (at 25°C)
96% min by GC
CH3(CH2)5CH3
48mbar at 20°C
1206
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CCCCCCC
100.21
CHEBI:43098
≥96%
Heptane
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!

DOTInformation : DOT Class 3, : Flammable Liquid

SDS
missing translation for 'documents'

missing translation for 'documents'