missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptane (Technical), Fisher Chemical™

Supplier:  Fisher Chemical BPH20200

 View more versions of this product

Catalog No. H20-200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Heptane
142-82-5
0.683g/cm³
200 L
Pass Test
Liquid
MFCD00009544
0.4 mPa/s at 20°C
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CCCCCCC
100.21
CHEBI:43098
Technical
-91°C
Colorless
98°C
−4°C (25°F)
Steel Drum
C7H16
48mbar at 20°C
1206
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.2

Chemical Identifiers

142-82-5
100.21
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
C7H16
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Accessories
Promotions

Promotions