Learn More
Heptanoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 164170025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Heptanoic acid | |
111-14-8 | |
White | |
223.0°C to 223.4°C | |
Authentic | |
Glass bottle | |
1.4210 to 1.4240 | |
MFCD00004426 | |
02,338 | |
15,4695 | |
Solubility in water: 0.24g/100mL (15°C). Other solubilities: soluble in alcohol, ether, dmf, chloroform, petrol, soluble in most organic solvents | |
CCCCCCC(=O)O | |
130.19 | |
3.40 mPa.s (30°C) | |
130.19 | |
Powder |
98% | |
-10.5°C | |
0.9100g/mL | |
>112°C | |
98% | |
C7H14O2 | |
CH3(CH2)5CO2H | |
2.5 L | |
0.91 | |
enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid | |
MNWFXJYAOYHMED-UHFFFAOYSA-N | |
heptanoic acid | |
8094 | |
CHEBI:45571 | |
98% |
Chemical Identifiers
111-14-8 | |
130.19 | |
MNWFXJYAOYHMED-UHFFFAOYSA-N | |
8094 | |
heptanoic acid |
C7H14O2 | |
MFCD00004426 | |
enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid | |
CHEBI:45571 | |
CCCCCCC(=O)O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 203-838-7
RTECSNumber : MJ1575000
TSCA : TSCA
RUO – Research Use Only