missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptanoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 164172500

 View more versions of this product

Catalog No. AC164172500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Heptanoic acid
111-14-8
White
223.0°C to 223.4°C
Authentic
Glass bottle
1.4210 to 1.4240
MFCD00004426
02,338
15,4695
Solubility in water: 0.24g/100mL (15°C). Other solubilities: soluble in alcohol, ether, dmf, chloroform, petrol, soluble in most organic solvents
CCCCCCC(=O)O
130.19
3.40 mPa.s (30°C)
130.19
Powder
98%
-10.5°C
0.9100g/mL
>112°C
98%
C7H14O2
CH3(CH2)5CO2H
250 mL
0.91
enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
MNWFXJYAOYHMED-UHFFFAOYSA-N
heptanoic acid
8094
CHEBI:45571
98%

Chemical Identifiers

111-14-8
130.19
MNWFXJYAOYHMED-UHFFFAOYSA-N
8094
heptanoic acid
C7H14O2
MFCD00004426
enanthic acid, oenanthic acid, n-heptanoic acid, enanthylic acid, heptoic acid, n-heptoic acid, n-heptylic acid, heptylic acid, oenanthylic acid, 1-hexanecarboxylic acid
CHEBI:45571
CCCCCCC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 203-838-7

RTECSNumber : MJ1575000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only