Learn More
Heptanoyl chloride, 99%
CAS: 2528-61-2 | C7H13ClO | 148.63 g/mol
$84.38 - $317.75
Chemical Identifiers
| CAS | 2528-61-2 |
|---|---|
| Molecular Formula | C7H13ClO |
| Molecular Weight (g/mol) | 148.63 |
| MDL Number | MFCD00000763 |
| InChI Key | UCVODTZQZHMTPN-UHFFFAOYSA-N |
| Synonym | enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride |
| PubChem CID | 17313 |
| IUPAC Name | heptanoyl chloride |
| SMILES | CCCCCCC(Cl)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0331518
|
Thermo Scientific Chemicals
L0331518 |
50 g |
Each for $84.38
|
|
|||||
|
AAL0331530
|
Thermo Scientific Chemicals
L0331530 |
250 g |
Each for $317.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2528-61-2 | |
| 148.63 | |
| UCVODTZQZHMTPN-UHFFFAOYSA-N | |
| 17313 | |
| CCCCCCC(Cl)=O |
| C7H13ClO | |
| MFCD00000763 | |
| enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride | |
| heptanoyl chloride |
Specifications
| 2528-61-2 | |
| 0.96 | |
| 58°C (136°F) | |
| C7H13ClO | |
| CH3(CH2)5COCl | |
| 50 g | |
| 605636 | |
| enanthyl chloride, n-heptanoyl chloride, enanthic chloride, oenanthic chloride, heptanoic acid chloride, heptanoylchloride, enanthylchloride, enanthoyl chloride, c6h13cocl, heptanoyl chloride | |
| CCCCCCC(Cl)=O | |
| 148.63 | |
| 148.63 | |
| Heptanoyl chloride |
| -84°C | |
| 172°C to 174°C | |
| Acrid | |
| 1.431 | |
| MFCD00000763 | |
| UN2920 | |
| Moisture sensitive | |
| UCVODTZQZHMTPN-UHFFFAOYSA-N | |
| heptanoyl chloride | |
| 17313 | |
| 99% |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P234-P235-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c
H227-H290-H314-H330
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 219-775-3
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only