missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Heptasaccharide Glc4Xyl3 80.0+%, TCI America™

Supplier:  TCI America H1044100MG

Catalog No. H1044100MG


Explore available promotions

Description

Description

Specifications

Specifications

Heptasaccharide Glc4Xyl3
White
MFCD04040861
XXXG, Xyloglucan Heptasaccharide
C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)O)CO)COC6C(C(C(CO6)O)O)O)COC7C(C(C(CO7)O)O)O)O)O)O)O)O)O
1062.924
1062.92
Crystalline Powder
121591-98-8
C39H66O33
100 mg
PZUPAGRIHCRVKN-UHFFFAOYSA-N
5-[5-[3,4-dihydroxy-6-[(3,4,5-trihydroxyoxan-2-yl)oxymethyl]-5-[3,4,5-trihydroxy-6-[(3,4,5-trihydroxyoxan-2-yl)oxymethyl]oxan-2-yl]oxyoxan-2-yl]oxy-3,4-dihydroxy-6-[(3,4,5-trihydroxyoxan-2-yl)oxymethyl]oxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,4-triol
44630346
≥93.0% (HPLC)

Chemical Identifiers

121591-98-8
1062.924
PZUPAGRIHCRVKN-UHFFFAOYSA-N
44630346
C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)O)CO)COC6C(C(C(CO6)O)O)O)COC7C(C(C(CO7)O)O)O)O)O)O)O)O)O
C39H66O33
MFCD04040861
XXXG, Xyloglucan Heptasaccharide
5-[5-[3,4-dihydroxy-6-[(3,4,5-trihydroxyoxan-2-yl)oxymethyl]-5-[3,4,5-trihydroxy-6-[(3,4,5-trihydroxyoxan-2-yl)oxymethyl]oxan-2-yl]oxyoxan-2-yl]oxy-3,4-dihydroxy-6-[(3,4,5-trihydroxyoxan-2-yl)oxymethyl]oxan-2-yl]oxy-6-(hydroxymethyl)oxane-2,3,4-triol
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions