missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Heptyl Chloroformate 98.0+%, TCI America™
Supplier: TCI America C072225ML
Specifications
Heptyl Chloroformate | |
Colorless | |
C8H15ClO2 | |
25 mL | |
Chloroformic Acid Heptyl Ester | |
CCCCCCCOC(=O)Cl | |
178.656 | |
178.66 | |
Liquid |
33758-34-8 | |
71°C | |
MFCD00059484 | |
3277 | |
SATRZZYUXUGZIE-UHFFFAOYSA-N | |
heptyl carbonochloridate | |
118547 | |
≥98.0% (GC,T) |
Chemical Identifiers
33758-34-8 | |
178.656 | |
SATRZZYUXUGZIE-UHFFFAOYSA-N | |
118547 | |
CCCCCCCOC(=O)Cl |
C8H15ClO2 | |
MFCD00059484 | |
Chloroformic Acid Heptyl Ester | |
heptyl carbonochloridate |
Safety and Handling
TSCA : Yes