missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Hesperidin | |
254°C | |
C28H34O15 | |
100 g | |
QUQPHWDTPGMPEX-QJBIFVCTSA-N | |
(2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one | |
45358131 | |
≥90.0% (HPLC,T) |
520-26-3 | |
Yellow | |
MFCD00075663 | |
Vitamin P | |
COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1 | |
610.57 | |
610.57 | |
Crystalline Powder |
Chemical Identifiers
520-26-3 | |
610.57 | |
QUQPHWDTPGMPEX-QJBIFVCTSA-N | |
45358131 | |
COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1 |
C28H34O15 | |
MFCD00075663 | |
Vitamin P | |
(2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one |
Safety and Handling
RTECSNumber : MK6650000
TSCA : Yes