Learn More
Hexaammineruthenium(III) chloride, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 363340010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Hexaammineruthenium(III) chloride | |
14282-91-8 | |
1 g | |
Cl3H18N6Ru | |
hexaammineruthenium iii chloride, hexaammineruthenium trichloride, hexaamminetrichlororuthenium, hexaammineruthenium trichloride, hydrate, ruthenium 3+ , hexaammine-, trichloride, hydrate, ruthenium 3+ chloride ammoniate 1:3:6, hexaammineruthenium iii trichloride, ruthenium 3+ ion hexaamine trichloride, ruthenium 3+ , hexaammine-, trichloride | |
GBDZMMXUOBAJMN-UHFFFAOYSA-K | |
azane;ruthenium(3+);trichloride | |
159731 | |
98% |
Yellow | |
Crystalline Powder | |
98% | |
MFCD00011478 | |
Solubility in water: soluble | |
N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3] | |
309.61 | |
309.61 | |
Glass bottle |
Chemical Identifiers
14282-91-8 | |
309.61 | |
GBDZMMXUOBAJMN-UHFFFAOYSA-K | |
159731 |
Cl3H18N6Ru | |
MFCD00011478 | |
hexaammineruthenium iii chloride, hexaammineruthenium trichloride, hexaamminetrichlororuthenium, hexaammineruthenium trichloride, hydrate, ruthenium 3+ , hexaammine-, trichloride, hydrate, ruthenium 3+ chloride ammoniate 1:3:6, hexaammineruthenium iii trichloride, ruthenium 3+ ion hexaamine trichloride, ruthenium 3+ , hexaammine-, trichloride | |
N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Ru+3] |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 238-176-8
RUO – Research Use Only