missing translation for 'onlineSavingsMsg'
Learn More

Hexachlorodisilane 98.0+%, TCI America™

$221.83 - $661.75

Chemical Identifiers

CAS 13465-77-5
Molecular Formula Cl6Si2
Molecular Weight (g/mol) 268.87
MDL Number MFCD00011599
InChI Key LXEXBJXDGVGRAR-UHFFFAOYSA-N
PubChem CID 83497
IUPAC Name trichloro(trichlorosilyl)silane
SMILES [Si]([Si](Cl)(Cl)Cl)(Cl)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H10285G
View Documents
TCI America
H10285G
5 g
Each for $221.83
Only null left
 
H102825G
View Documents
TCI America
H102825G
25 g
Each for $661.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13465-77-5
268.87
LXEXBJXDGVGRAR-UHFFFAOYSA-N
trichloro(trichlorosilyl)silane
Cl6Si2
MFCD00011599
83497
[Si]([Si](Cl)(Cl)Cl)(Cl)(Cl)Cl

Specifications

13465-77-5
Cl6Si2
5 g
LXEXBJXDGVGRAR-UHFFFAOYSA-N
trichloro(trichlorosilyl)silane
83497
≥98.0% (T)
Hexachlorodisilane
Colorless
MFCD00011599
2987
[Si]([Si](Cl)(Cl)Cl)(Cl)(Cl)Cl
268.87
268.87
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (1)-1214

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'