missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexadecanamide 95.0+%, TCI America™
$276.81 - $558.83
Chemical Identifiers
CAS | 629-54-9 |
---|---|
Molecular Formula | C16H33NO |
Molecular Weight (g/mol) | 255.446 |
MDL Number | MFCD00025534 |
InChI Key | HSEMFIZWXHQJAE-UHFFFAOYSA-N |
Synonym | Palmitamide |
PubChem CID | 69421 |
ChEBI | CHEBI:74475 |
IUPAC Name | hexadecanamide |
SMILES | CCCCCCCCCCCCCCCC(=O)N |
Chemical Identifiers
629-54-9 | |
255.446 | |
HSEMFIZWXHQJAE-UHFFFAOYSA-N | |
69421 | |
hexadecanamide |
C16H33NO | |
MFCD00025534 | |
Palmitamide | |
CHEBI:74475 | |
CCCCCCCCCCCCCCCC(=O)N |
Specifications
629-54-9 | |
White | |
C16H33NO | |
10 g | |
HSEMFIZWXHQJAE-UHFFFAOYSA-N | |
hexadecanamide | |
69421 | |
255.45 | |
Crystalline Powder |
106°C | |
236°C | |
MFCD00025534 | |
Palmitamide | |
CCCCCCCCCCCCCCCC(=O)N | |
255.446 | |
CHEBI:74475 | |
≥95.0% (GC) | |
Hexadecanamide |
Safety and Handling
EINECSNumber : (2)-0824
TSCA : Yes