missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecylphosphonic Acid 98.0+%, TCI America™
$193.86 - $674.79
Chemical Identifiers
| CAS | 4721-17-9 |
|---|---|
| Molecular Formula | C16H35O3P |
| Molecular Weight (g/mol) | 306.427 |
| MDL Number | MFCD00015836 |
| InChI Key | JDPSFRXPDJVJMV-UHFFFAOYSA-N |
| PubChem CID | 13783525 |
| IUPAC Name | hexadecylphosphonic acid |
| SMILES | CCCCCCCCCCCCCCCCP(=O)(O)O |
Chemical Identifiers
| 4721-17-9 | |
| 306.427 | |
| JDPSFRXPDJVJMV-UHFFFAOYSA-N | |
| hexadecylphosphonic acid |
| C16H35O3P | |
| MFCD00015836 | |
| 13783525 | |
| CCCCCCCCCCCCCCCCP(=O)(O)O |
Specifications
| 4721-17-9 | |
| White | |
| MFCD00015836 | |
| JDPSFRXPDJVJMV-UHFFFAOYSA-N | |
| hexadecylphosphonic acid | |
| 13783525 | |
| ≥98.0% (T) | |
| Hexadecylphosphonic Acid |
| 95°C | |
| C16H35O3P | |
| 1 g | |
| CCCCCCCCCCCCCCCCP(=O)(O)O | |
| 306.427 | |
| 306.43 | |
| Crystalline Powder |
Safety and Handling
TSCA : No