missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexadecyltrimethylammonium bromide, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 227160250

 View more versions of this product

Catalog No. AC227160250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Hexadecyltrimethylammonium Bromide
57-09-0
230.0°C
25 g
Authentic
99+%
CH3(CH2)15N(CH3)3Br
04, IV, 819
15, 2024
Solubility in water: 13g/L (20°C). Other solubilities: soluble in methanol (>200g/L at 20 °C), ethanol, insoluble in benzene, diethyl ether, chloroform, sparingly soluble in acetone
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
364.46
CHEBI:3567
≥99%
White
Crystalline Powder
>230°C
244°C
Plastic Bottle
C19H42BrN
MFCD00011772
15,77
Cetrimonium Bromide, Cetyltrimethylammonium Bromide, CTABr
LZZYPRNAOMGNLH-UHFFFAOYSA-M
hexadecyltrimethylazanium bromide
5974
364.45

Chemical Identifiers

57-09-0
364.46
LZZYPRNAOMGNLH-UHFFFAOYSA-M
CHEBI:3567
C19H42BrN
MFCD00011772
5974
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
Safety and Handling

Safety and Handling

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only