missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexadecyltrimethylammonium bromide, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 227161000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Hexadecyltrimethylammonium Bromide | |
57-09-0 | |
230.0°C | |
99+% | |
CH3(CH2)15N(CH3)3Br | |
100 g | |
15,77 | |
Cetrimonium Bromide, Cetyltrimethylammonium Bromide, CTABr | |
244°C | |
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C | |
364.46 | |
CHEBI:3567 | |
≥99% | |
Plastic Bottle |
White | |
Crystalline Powder | |
>230°C | |
C19H42BrN | |
MFCD00011772 | |
04, IV, 819 | |
15, 2024 | |
Solubility in water: 13g/L (20°C). Other solubilities: soluble in methanol (>200g/L at 20 °C), ethanol, insoluble in benzene, diethyl ether, chloroform, sparingly soluble in acetone | |
LZZYPRNAOMGNLH-UHFFFAOYSA-M | |
hexadecyltrimethylazanium bromide | |
5974 | |
364.45 | |
Authentic |
Chemical Identifiers
57-09-0 | |
364.46 | |
LZZYPRNAOMGNLH-UHFFFAOYSA-M | |
CHEBI:3567 |
C19H42BrN | |
MFCD00011772 | |
5974 | |
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C |
Safety and Handling
Warning
RUO – Research Use Only