missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexadecyltrimethylammonium Perchlorate 98.0+%, TCI America™
Supplier: TCI America H07355G
Specifications
Hexadecyltrimethylammonium Perchlorate | |
White | |
MFCD00144895 | |
1479 | |
KXNZYPBRSATHKV-UHFFFAOYSA-M | |
hexadecyltrimethylazanium perchlorate | |
15520692 | |
≥98.0% (N) |
6941-37-3 | |
C19H42ClNO4 | |
5 g | |
Cetyltrimethylammonium Perchlorate | |
[O-][Cl](=O)(=O)=O.CCCCCCCCCCCCCCCC[N+](C)(C)C | |
384.00 | |
384.00 | |
Crystalline Powder |
Chemical Identifiers
6941-37-3 | |
384.00 | |
KXNZYPBRSATHKV-UHFFFAOYSA-M | |
15520692 | |
[O-][Cl](=O)(=O)=O.CCCCCCCCCCCCCCCC[N+](C)(C)C |
C19H42ClNO4 | |
MFCD00144895 | |
Cetyltrimethylammonium Perchlorate | |
hexadecyltrimethylazanium perchlorate |
Safety and Handling
EINECSNumber : (2)-0184&(9)-0795&(9)-1971
TSCA : No