missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexafluoroisopropyl Methyl Ether 98.0+%, TCI America™
$170.11 - $510.32
Chemical Identifiers
CAS | 13171-18-1 |
---|---|
Molecular Formula | C4H4F6O |
Molecular Weight (g/mol) | 182.065 |
MDL Number | MFCD00221772 |
InChI Key | VNXYDFNVQBICRO-UHFFFAOYSA-N |
Synonym | 1,1,1,3,3,3-Hexafluoro-2-methoxypropane, Isoindoklon |
PubChem CID | 25749 |
IUPAC Name | 1,1,1,3,3,3-hexafluoro-2-methoxypropane |
SMILES | COC(C(F)(F)F)C(F)(F)F |
Chemical Identifiers
13171-18-1 | |
182.065 | |
VNXYDFNVQBICRO-UHFFFAOYSA-N | |
25749 | |
COC(C(F)(F)F)C(F)(F)F |
C4H4F6O | |
MFCD00221772 | |
1,1,1,3,3,3-Hexafluoro-2-methoxypropane, Isoindoklon | |
1,1,1,3,3,3-hexafluoro-2-methoxypropane |
Specifications
13171-18-1 | |
50°C | |
MFCD00221772 | |
3271 | |
VNXYDFNVQBICRO-UHFFFAOYSA-N | |
1,1,1,3,3,3-hexafluoro-2-methoxypropane | |
25749 | |
≥98.0% (GC) | |
Hexafluoroisopropyl Methyl Ether |
Colorless | |
C4H4F6O | |
5 g | |
1,1,1,3,3,3-Hexafluoro-2-methoxypropane, Isoindoklon | |
COC(C(F)(F)F)C(F)(F)F | |
182.065 | |
182.07 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-4050
RTECSNumber : KO2282000
TSCA : No