Learn More
Hexafluorophosphoric acid, 60 wt.% solution in water
CAS: 16940-81-1 | HF6P
$40.94 - $212.55
Chemical Identifiers
| CAS | 16940-81-1 |
|---|---|
| Molecular Formula | HF6P |
| MDL Number | MFCD00011350 |
| Synonym | Hydrogen hexafluorophosphate |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC191110250
|
Thermo Scientific Chemicals
191110250 |
25 g | Plastic bottle |
Each for $40.94
|
|
||||
|
AC191115000
|
Thermo Scientific Chemicals
191115000 |
500 g | Plastic bottle |
Each for $212.55
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 16940-81-1 | |
| HF6P | |
| MFCD00011350 | |
| Hydrogen hexafluorophosphate |
Specifications
| Colorless | |
| HF6P | |
| MFCD00011350 | |
| 01,42; 02,402 | |
| Hydrogen hexafluorophosphate | |
| 145.97 | |
| Plastic bottle | |
| 25 g | |
| Hexafluorophosphoric acid, 60 wt% Solution in Water |
| 16940-81-1,7664-38-2,7664-39-3,13537-32-1,13779-41-4,7732-18-5 | |
| HPF6 | |
| contains the following products of hydrolysis: HPO2F2, H2PO3F, H3PO4 and HF. the presence of these hydrolysis products may rise the strength to over 70% | |
| 1.7500g/mL | |
| Solubility in water: hydrolyzes slowly. | |
| ≥60% (PF6-content) | |
| 1.75 | |
| Fuming Solution |
Safety and Handling
GHS H Statement:
Fatal in contact with skin.
Causes serious eye damage.
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 241-006-5
RTECSNumber : SY7115000
TSCA : TSCA
RUO – Research Use Only