Learn More
Hexamethyldisilane, 98+%
CAS: 1450-14-2 | C6H18Si2 | 146.38 g/mol
$101.87 - $327.72
Chemical Identifiers
| CAS | 1450-14-2 |
|---|---|
| Molecular Formula | C6H18Si2 |
| Molecular Weight (g/mol) | 146.38 |
| MDL Number | MFCD00008258 |
| InChI Key | NEXSMEBSBIABKL-UHFFFAOYSA-N |
| Synonym | hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane |
| PubChem CID | 74057 |
| IUPAC Name | trimethyl(trimethylsilyl)silane |
| SMILES | C[Si](C)(C)[Si](C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC202690100
|
Thermo Scientific Chemicals
202690100 |
10 mL | Glass bottle |
Each for $101.87
|
|
||||
|
AC202690500
|
Thermo Scientific Chemicals
202690500 |
50 mL | Glass bottle |
Each for $327.72
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1450-14-2 | |
| 146.38 | |
| NEXSMEBSBIABKL-UHFFFAOYSA-N | |
| 74057 | |
| C[Si](C)(C)[Si](C)(C)C |
| C6H18Si2 | |
| MFCD00008258 | |
| hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane | |
| trimethyl(trimethylsilyl)silane |
Specifications
| 1450-14-2 | |
| Colorless | |
| 112°C to 114°C | |
| Authentic | |
| Glass bottle | |
| 1.4211 to 1.4231 | |
| MFCD00008258 | |
| 04, IV, 4277 | |
| 0.71 | |
| NEXSMEBSBIABKL-UHFFFAOYSA-N | |
| trimethyl(trimethylsilyl)silane | |
| 74057 | |
| 98+% | |
| Hexamethyldisilane |
| 15°C | |
| 0.7100g/mL | |
| 11°C | |
| 98+% | |
| C6H18Si2 | |
| (CH3)3SiSi(CH3)3 | |
| 10 mL | |
| 11,253; 15,164 | |
| hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane | |
| C[Si](C)(C)[Si](C)(C)C | |
| 146.38 | |
| 146.38 | |
| Liquid |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 215-911-
RUO – Research Use Only