missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethyldisilane 98.0+%, TCI America™
$65.24 - $370.95
Chemical Identifiers
| CAS | 1450-14-2 |
|---|---|
| Molecular Formula | C6H18Si2 |
| Molecular Weight (g/mol) | 146.38 |
| MDL Number | MFCD00008258 |
| InChI Key | NEXSMEBSBIABKL-UHFFFAOYSA-N |
| Synonym | hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane |
| PubChem CID | 74057 |
| IUPAC Name | hexamethyldisilane |
| SMILES | C[Si](C)(C)[Si](C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H063810ML
|
TCI America
H063810ML |
10 mL |
Each for $65.24
|
|
|||||
|
H0638100ML
|
TCI America
H0638100ML |
100 mL |
Each for $370.95
|
|
|||||
Chemical Identifiers
| 1450-14-2 | |
| 146.38 | |
| NEXSMEBSBIABKL-UHFFFAOYSA-N | |
| 74057 | |
| C[Si](C)(C)[Si](C)(C)C |
| C6H18Si2 | |
| MFCD00008258 | |
| hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane | |
| hexamethyldisilane |
Specifications
| 1450-14-2 | |
| 113°C | |
| MFCD00008258 | |
| 1993 | |
| NEXSMEBSBIABKL-UHFFFAOYSA-N | |
| hexamethyldisilane | |
| 74057 | |
| ≥98.0% (GC) | |
| Hexamethyldisilane |
| Yellow | |
| C6H18Si2 | |
| 10 mL | |
| hexamethyldisilane, disilane, hexamethyl, permethyldisilane, disilane, 1,1,1,2,2,2-hexamethyl, 1,1,1,2,2,2-hexamethyldisilane, trimethyl trimethylsilyl silane, ch3 6si2, hecamethyldisilane, disilane m6, hexamethyl disilane | |
| C[Si](C)(C)[Si](C)(C)C | |
| 146.38 | |
| 146.38 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-3711
RTECSNumber : JM9170000
TSCA : Yes