missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexamethyldisilazane, Electronic grade, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 4203922

 View more versions of this product

Catalog No. AA4203922


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Hexamethyldisilazane
-78°C
0.774 g/mL
8°C (46°F)
C6H19NSi2
(CH3)3SiNHSi(CH3)3
100 g
635752
14,4689
Miscible with acetone,benzene,ethyl ether,heptane and perchloroethylene.
C[Si](C)(C)N[Si](C)(C)C
161.395
CHEBI:85068
≥99%
Liquid
999-97-3
Colorless
126°C
Odorless
1.408
MFCD00008259
UN3286
Moisture sensitive
hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
FFUAGWLWBBFQJT-UHFFFAOYSA-N
[dimethyl-(trimethylsilylamino)silyl]methane
13838
161.4
Electronic

Chemical Identifiers

999-97-3
161.395
FFUAGWLWBBFQJT-UHFFFAOYSA-N
13838
[dimethyl-(trimethylsilylamino)silyl]methane
C6H19NSi2
MFCD00008259
hexamethyldisilazane, bis trimethylsilyl amine, hmds, 1,1,1,3,3,3-hexamethyldisilazane, hexamethylsilazane, silanamine, 1,1,1-trimethyl-n-trimethylsilyl, tri-sil, 1,1,1-trimethyl-n-trimethylsilyl silanamine, hexamethyldisilizane, disilazane, 1,1,1,3,3,3-hexamethyl
CHEBI:85068
C[Si](C)(C)N[Si](C)(C)C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H311-H331-H314-H318-H302
Highly flammable liquid and vapor.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H332-H311-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.

EINECSNumber : 213-668-5

RTECSNumber : JM9230000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions