Learn More
Hexamethyldisiloxane, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A11848AP
Description
Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule. It is also used to make low-k dielectric materials for the semi-conductor industries by plasma-enhanced chemical vapor deposition(PECVD) method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule. It is also used to make low-k dielectric materials for the semi-conductor industries by plasma-enhanced chemical vapor deposition(PECVD) method.
Solubility
Slightly miscible with water.
Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases and oxygen.
Specifications
Hexamethyldisiloxane | |
-67°C | |
100°C to 101°C | |
Characteristic | |
1.377 | |
MFCD00008265 | |
UN1993 | |
hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide | |
UQEAIHBTYFGYIE-UHFFFAOYSA-N | |
trimethyl(trimethylsilyloxy)silane | |
24764 | |
162.38 |
107-46-0 | |
0.764 | |
−2°C (28°F) | |
C6H18OSi2 | |
(CH3)3SiOSi(CH3)3 | |
500 mL | |
1736258 | |
Slightly miscible with water. | |
C[Si](C)(C)O[Si](C)(C)C | |
162.379 | |
CHEBI:78002 | |
≥98% |
Chemical Identifiers
107-46-0 | |
MFCD00008265 | |
hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide | |
CHEBI:78002 | |
C[Si](C)(C)O[Si](C)(C)C |
C6H18OSi2 | |
UQEAIHBTYFGYIE-UHFFFAOYSA-N | |
24764 | |
trimethyl(trimethylsilyloxy)silane |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P271-P280-P303+P361+P353-P304+P340-P312-P314-P370+P378q-P501c
H225
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 203-492-7
RTECSNumber : JM9237000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only