Learn More
Hexamethylenetetramine, 98.5%, contains an anticaking agent, Thermo Scientific Chemicals
$88.49 - $167.24
Chemical Identifiers
CAS | 100-97-0 |
---|---|
Molecular Formula | C6H12N4 |
Molecular Weight (g/mol) | 140.19 |
MDL Number | MFCD00006895 |
InChI Key | VKYKSIONXSXAKP-UHFFFAOYSA-N |
Synonym | methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine |
PubChem CID | 4101 |
ChEBI | CHEBI:6824 |
SMILES | C1N2CN3CN1CN(C2)C3 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC232890010
|
Thermo Scientific Chemicals
232890010 |
1 kg | Plastic bottle |
Each for $88.49
|
|
||||
AC232890025
|
Thermo Scientific Chemicals
232890025 |
2.5 kg | Plastic bottle |
Each for $167.24
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-97-0 | |
140.19 | |
VKYKSIONXSXAKP-UHFFFAOYSA-N | |
4101 | |
C1N2CN3CN1CN(C2)C3 |
C6H12N4 | |
MFCD00006895 | |
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine | |
CHEBI:6824 |
Specifications
100-97-0 | |
White | |
250°C | |
98.5% | |
C6H12N4 | |
1 kg | |
01,427; 02,208; 04,243; 09,234 | |
15, 6038 | |
263.0 °C | |
VKYKSIONXSXAKP-UHFFFAOYSA-N | |
140.19 | |
CHEBI:6824 | |
98.5% | |
Hexamethylenetetramine, 98.5% |
260.0°C | |
1.3300g/mL | |
Authentic | |
Plastic bottle | |
MFCD00006895 | |
01, 583 | |
1.33 | |
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine | |
Solubility in water: 895g/L (20°C). Other solubilities: soluble in chloroform: 13.4% (20°C), soluble in ethanol: 2.89% (20°C), soluble in acetone: 0.65% (20°C), insoluble in ether: 0.06% (20°C), soluble in methanol: 7.25% (20°C) | |
C1N2CN3CN1CN(C2)C3 | |
4101 | |
140.19 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Flammable solid.
GHS P Statement
Wear protective gloves/protective clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
EINECSNumber : 202-905-8
RTECSNumber : MN4725000
TSCA : TSCA
RUO – Research Use Only