missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexamethylenetetramine, ACS, 99%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor M121712KG
Specifications
| 100-97-0 | |
| 0.001 | |
| Poly Pail | |
| 12 kg | |
| C1N2CN3CN1CN(C2)C3 | |
| 140.19 | |
| ACS |
| 1 | |
| 0.02 | |
| C6H12N4 | |
| VKYKSIONXSXAKP-UHFFFAOYSA-N | |
| 1,3,5,7-tetraazatricyclo[3.3.1.1³,â·]decane | |
| 0.99 |
Chemical Identifiers
| 100-97-0 | |
| 140.19 | |
| 1,3,5,7-tetraazatricyclo[3.3.1.1³,â·]decane |
| C6H12N4 | |
| VKYKSIONXSXAKP-UHFFFAOYSA-N | |
| C1N2CN3CN1CN(C2)C3 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.