Learn More
Hexamethylenetetramine, ACS, 99+%, Thermo Scientific Chemicals
$50.14 - $336.07
Chemical Identifiers
CAS | 100-97-0 |
---|---|
Molecular Formula | C6H12N4 |
Molecular Weight (g/mol) | 140.19 |
MDL Number | MFCD00006895 |
InChI Key | VKYKSIONXSXAKP-UHFFFAOYSA-N |
Synonym | methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine |
PubChem CID | 4101 |
ChEBI | CHEBI:6824 |
SMILES | C1N2CN3CN1CN(C2)C3 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA3646222
|
Thermo Scientific Chemicals
03646222 |
100 g |
Each for $50.14
|
|
|||||
AA3646236
|
Thermo Scientific Chemicals
03646236 |
500 g |
Each for $114.67
|
|
|||||
AA36462A3
|
Thermo Scientific Chemicals
036462A3 |
2 kg |
Each for $336.07
|
|
|||||
Description
Hexamethylenetetramine is used in the manufacturing of phenolic resins liquid preparations and molding compounds. It finds applications in in brake and clutch linings, abrasive products, non-woven textiles and fireproof materials. It is also used as a drug taken by mouth to prevent recurrent urinary tract infections. It is used as a solid fuel in association with 1,3,5-trioxane in campers, hobbyists, the military and relief organizations. It acts as a food additive and as a preservative. It is an important reagent in organic synthesis, especially Duff reaction, Sommelet reaction and in the Delepine reaction. Further, it is used in adhesives, coatings, sealants, and leather preservatives. It is utilized as a formaldehyde releaser for photographic film development and as a hardener for epoxy resins of the bisphenol A type.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-97-0 | |
140.19 | |
VKYKSIONXSXAKP-UHFFFAOYSA-N | |
4101 | |
C1N2CN3CN1CN(C2)C3 |
C6H12N4 | |
MFCD00006895 | |
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine | |
CHEBI:6824 |
Specifications
100-97-0 | |
White | |
250°C (482°F) | |
MFCD00006895 | |
UN1328 | |
Hygroscopic | |
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine | |
VKYKSIONXSXAKP-UHFFFAOYSA-N | |
140.19 | |
CHEBI:6824 | |
≥99% | |
Solid |
280°C (sublimation) | |
1.33 g/mL | |
C6H12N4 | |
100 g | |
2018 | |
14,5966 | |
Soluble in water,dimethyl sulfoxide,chloroform,ethanol and acetone. Slightly soluble in ether. | |
C1N2CN3CN1CN(C2)C3 | |
4101 | |
140.19 | |
ACS | |
Hexamethylenetetramine |
Safety and Handling
GHS H Statement
H228-H317
Flammable solid.
May cause an allergic skin reaction.
P210-P240-P241-P261-P272-P280-P302+P352-P333+P313-P363-P370+P378q-P501c
H228-H317-H500
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: HEXAMETHYLENETETRAMINE
EINECSNumber : 202-905-8
RTECSNumber : MN4725000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only