missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexamethylenetetramine (Certified ACS), Fisher Chemical
Supplier: Fisher Chemical H288500
Specifications
Hexamethylenetetramine | |
263°C (sublimes) | |
1.330g/cm³ | |
0.1% max. | |
≥99.0% | |
C6H12N4 | |
500 g | |
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine | |
C1N2CN3CN1CN(C2)C3 | |
4101 | |
140.19 | |
Certified ACS |
100-97-0 | |
White | |
8.5 to 9.5 | |
2.0% max. | |
Glass Bottle | |
MFCD00006895 | |
1328 | |
VKYKSIONXSXAKP-UHFFFAOYSA-N | |
140.19 | |
CHEBI:6824 | |
≥99.0% | |
Solid |
Chemical Identifiers
100-97-0 | |
140.19 | |
VKYKSIONXSXAKP-UHFFFAOYSA-N | |
4101 | |
C1N2CN3CN1CN(C2)C3 |
C6H12N4 | |
MFCD00006895 | |
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine | |
CHEBI:6824 |
Safety and Handling
Emergency Overview
Flammable solid. May cause an allergic skin reaction.
NFPA
Health: 2
Flammability:
Signal Word: WARNING!
DOTInformation : Class 4.1: Flammable Solid