missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexamethylenetetramine (Crystalline/USP), Fisher Chemical

Supplier:  Thermo Fisher Scientific H290500

Catalog No. H290-500



Explore available promotions

Description

Description

Specifications

Specifications

Hexamethylenetetramine
White
8.5 to 9.5
2.0% max.
Glass Bottle
MFCD00006895
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
VKYKSIONXSXAKP-UHFFFAOYSA-N
140.19
CHEBI:6824
99.0 to 100.5%
Solid
100-97-0
1.330g/cm³
0.1% max.
99.0 to 100.5 %
C6H12N4
500 g
Meets Requirements
C1N2CN3CN1CN(C2)C3
4101
140.19
USP

Chemical Identifiers

100-97-0
140.19
VKYKSIONXSXAKP-UHFFFAOYSA-N
4101
C1N2CN3CN1CN(C2)C3
C6H12N4
MFCD00006895
methenamine, hexamethylenetetramine, hexamine, urotropine, 1,3,5,7-tetraazaadamantane, hexamethylenamine, aminoform, methenamin, urotropin, formamine
CHEBI:6824
Safety and Handling

Safety and Handling

Emergency Overview
Flammable solid. May cause an allergic skin reaction.
NFPA
Health: 2
Flammability:

Signal Word: WARNING!

SDS Documents

Documents

Promotions

Promotions