missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexamethylphosphoric Triamide 98.0+%, TCI America™
$47.40 - $292.30
Chemical Identifiers
CAS | 680-31-9 |
---|---|
Molecular Formula | C6H18N3OP |
Molecular Weight (g/mol) | 179.204 |
MDL Number | MFCD00008303 |
InChI Key | GNOIPBMMFNIUFM-UHFFFAOYSA-N |
Synonym | hexamethylphosphoramide, hmpa, hexametapol, hexamethylphosphoric triamide, hempa, hmpta, hexamethylphosphoric acid triamide, hexamethylphosphorictriamide, hexamethylphosphotriamide, phosphoric hexamethyltriamide |
PubChem CID | 12679 |
ChEBI | CHEBI:24565 |
IUPAC Name | N-[bis(dimethylamino)phosphoryl]-N-methylmethanamine |
SMILES | CN(C)P(=O)(N(C)C)N(C)C |
Chemical Identifiers
680-31-9 | |
179.204 | |
GNOIPBMMFNIUFM-UHFFFAOYSA-N | |
12679 | |
N-[bis(dimethylamino)phosphoryl]-N-methylmethanamine |
C6H18N3OP | |
MFCD00008303 | |
hexamethylphosphoramide, hmpa, hexametapol, hexamethylphosphoric triamide, hempa, hmpta, hexamethylphosphoric acid triamide, hexamethylphosphorictriamide, hexamethylphosphotriamide, phosphoric hexamethyltriamide | |
CHEBI:24565 | |
CN(C)P(=O)(N(C)C)N(C)C |
Specifications
680-31-9 | |
232°C | |
MFCD00008303 | |
hexamethylphosphoramide, hmpa, hexametapol, hexamethylphosphoric triamide, hempa, hmpta, hexamethylphosphoric acid triamide, hexamethylphosphorictriamide, hexamethylphosphotriamide, phosphoric hexamethyltriamide | |
CN(C)P(=O)(N(C)C)N(C)C | |
179.204 | |
CHEBI:24565 | |
≥98.0% (GC,T) | |
Hexamethylphosphoric Triamide |
Yellow | |
C6H18N3OP | |
25 g | |
GNOIPBMMFNIUFM-UHFFFAOYSA-N | |
N-[bis(dimethylamino)phosphoryl]-N-methylmethanamine | |
12679 | |
179.20 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-1930
RTECSNumber : TD0875000
TSCA : Yes