Learn More
Hexanal, 96%, stabilized, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 151850025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Hexanal | |
10mg KOH/g max. | |
130.0°C to 131.0°C | |
Authentic | |
Glass bottle | |
1.4020 to 1.4070 | |
MFCD00007027 | |
01, II, 745 | |
15, 1762 | |
Solubility in water: 4.8g/L (20°C). Other solubilities: soluble in methanol, acetone, chloroform, ether | |
CCCCCC=O | |
100.16 | |
6184 | |
100.16 |
66-25-1,59-02-9 | |
0.8150g/mL | |
32°C | |
95% min. (GC) | |
C6H12O | |
CH3(CH2)4CHO | |
2.5L | |
0.815 | |
caproaldehyde, hexaldehyde, caproic aldehyde, capronaldehyde, 1-hexanal, n-hexanal, hexanaldehyde, n-caproaldehyde, hexylaldehyde, aldehyde c-6 | |
JARKCYVAAOWBJS-UHFFFAOYSA-N | |
hexanal | |
approx. 500ppm alpha-tocopherol | |
0.69 mPa.s (20°C) | |
96% |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse caut
GHS Signal Word: Warning
EINECSNumber : 200-624-5
RUO â Research Use Only