missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexanamide 98.0+%, TCI America™
$130.48 - $337.40
Chemical Identifiers
| CAS | 628-02-4 |
|---|---|
| Molecular Formula | C6H13NO |
| Molecular Weight (g/mol) | 115.176 |
| MDL Number | MFCD00008046 |
| InChI Key | ALBYIUDWACNRRB-UHFFFAOYSA-N |
| Synonym | capronamide, hexamide, hexylamide, n-caproamide, caproamide, n-hexanamide, hexamide van, hexanoamide, n-capronamide, n-caproic amide |
| PubChem CID | 12332 |
| IUPAC Name | hexanamide |
| SMILES | CCCCCC(=O)N |
Chemical Identifiers
| 628-02-4 | |
| 115.176 | |
| ALBYIUDWACNRRB-UHFFFAOYSA-N | |
| 12332 | |
| CCCCCC(=O)N |
| C6H13NO | |
| MFCD00008046 | |
| capronamide, hexamide, hexylamide, n-caproamide, caproamide, n-hexanamide, hexamide van, hexanoamide, n-capronamide, n-caproic amide | |
| hexanamide |
Specifications
| 628-02-4 | |
| White | |
| MFCD00008046 | |
| capronamide, hexamide, hexylamide, n-caproamide, caproamide, n-hexanamide, hexamide van, hexanoamide, n-capronamide, n-caproic amide | |
| CCCCCC(=O)N | |
| 115.176 | |
| 115.18 | |
| Crystalline Powder |
| 100°C | |
| C6H13NO | |
| 5 g | |
| ALBYIUDWACNRRB-UHFFFAOYSA-N | |
| hexanamide | |
| 12332 | |
| ≥98.0% (N) | |
| Hexanamide |
Safety and Handling
EINECSNumber : (2)-0824
RTECSNumber : MN7875000
TSCA : Yes