missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, mixed isomers, ACS, 98.5+%

CAS: 92112-69-1 | C6H14 | 86.18 g/mol

$108.74 - $296.58

Chemical Identifiers

CAS 92112-69-1
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00009520
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA33321K2
View Documents
Thermo Scientific Chemicals
033321K2
1 L N/A
Only null left
 
AA33321K4
View Documents
Thermo Scientific Chemicals
033321K4
2.5 L N/A
Only null left
 
AA33321M1
View Documents
Thermo Scientific Chemicals
33321M1
4 x 1 L N/A
Only null left
 
AA33321K7
View Documents
Thermo Scientific Chemicals
033321K7
4 L N/A
Only null left
 
Description

Description

Hexane mixed isomers is used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds namely butyl lithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

92112-69-1
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD00009520
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC

Specifications

-95°C
Colorless
65°C to 70°C
<-23°C (-9°F)
1.3795
MFCD00009520
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
ACS
92112-69-1
0.67 g/mL
1 L
Liquid, Mixture Of 5 Isomers
C6H14
UN1208
Miscible with alcohol,chloroform,acetone and ether. Immiscible with water.
CCCCCC
86.18
CHEBI:29021
≥98.5%
Hexanes, Mixed isomers
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335-H336-H361f-H372

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES

EINECSNumber : 295-570-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only