HexArmor™ Arm Guards DFS Item
Reduce costs—last 10 to 15 times longer than typical sleeves
Supplier:  HexArmor™ AG10009SM
 View more versions of this product
Catalog No. 19-130-3752B
Explore available promotions
Description
Description
Provide a high level of protection to 9 in. of forearm from wrist up. Protects from metal, knives, and punctures.
  • Offer ISEA LEvel 5 cut resistance
  • Spandex™ wrist insert with thumb hole for comfortable fit
  • Will not slip down like knit sleeves
  • Compatible with most gloves
  • Ambidextrous
  • Machine washable
8 in. Arm Guards
  • Can be used in waste handling, recycling operations, lab work, and animal handling
  • 9 in. Arm Guards
    • Ideal for assembly, meat processing and packing; and wood, glass, and metal handling
    • Available in two styles: single-layer Arm Guard 9S and double-layer Arm Guard 9D which that enhanced puncture protection from needles, knives, and glass
    1These arm guards are cut and/or puncture resistant, not cut or puncture proof. Always conduct a safe test with your application to determine if the product is suited for your needs
    Specifications
    Specifications
    1-ply
    SuperFabric™
    Spandex™ wrist insert, thumb hole
    22.8 cm
    Medium
    Assembly, meat processing and packing, Wood, Glass and Metal handling
    9 in.