missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexyl Acetoacetate 97.0+%, TCI America™

Supplier:  TCI America A091325ML

Catalog No. A091325ML


Explore available promotions

Description

Description

Specifications

Specifications

Hexyl Acetoacetate
Yellow
C10H18O3
25 mL
QNZLAXONNWOLJY-UHFFFAOYSA-N
hexyl 3-oxobutanoate
83577
≥97.0% (GC)
13562-84-0
239°C
MFCD00059440
Acetoacetic Acid Hexyl Ester
CCCCCCOC(=O)CC(C)=O
186.25
186.25
Liquid

Chemical Identifiers

13562-84-0
186.25
QNZLAXONNWOLJY-UHFFFAOYSA-N
83577
CCCCCCOC(=O)CC(C)=O
C10H18O3
MFCD00059440
Acetoacetic Acid Hexyl Ester
hexyl 3-oxobutanoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1505

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions