missing translation for 'onlineSavingsMsg'
Learn More

Hexyl alcohol, 98%, pure

CAS: 111-27-3 | C6H14O | 102.18 g/mol

$59.81 - $1174.96

Chemical Identifiers

CAS 111-27-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00002982
InChI Key ZSIAUFGUXNUGDI-UHFFFAOYSA-N
Synonym 1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
PubChem CID 8103
ChEBI CHEBI:87393
IUPAC Name hexan-1-ol
SMILES CCCCCCO
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120791000
SDS View Documents
Thermo Scientific Chemicals
120791000
100 mL Glass bottle
Each for $59.81
Only null left
 
AC120790010
SDS View Documents
Thermo Scientific Chemicals
120790010
1 L Glass bottle
Each for $90.65
Case of 4 Each for $0.00
Only null left
 
AC120790025
SDS View Documents
Thermo Scientific Chemicals
120790025
2.5 L Glass bottle
Each for $186.31
Only null left
 
AC120790100
SDS View Documents
Thermo Scientific Chemicals
120790100
10 L Plastic drum
Each for $614.85
Only null left
 
AC120790250
SDS View Documents
Thermo Scientific Chemicals
120790250
25 L Metal Drum
Each for $1,174.96
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-27-3
102.18
ZSIAUFGUXNUGDI-UHFFFAOYSA-N
8103
hexan-1-ol
C6H14O
MFCD00002982
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
CHEBI:87393
CCCCCCO

Specifications

111-27-3
-52.0°C
156.0°C to 157.0°C
Authentic
Glass bottle
1.4170 to 1.4190
MFCD00002982
01, 407
15, 4734
Solubility in water: 6g/L (25°C). Other solubilities: miscible with alcohol and ether
CCCCCCO
102.18
5.3 mPa.s (20°C)
102.18
Pure
Hexyl alcohol
0.02mg KOH/g max.
0.8136g/mL
59°C
97.5% min. (GC)
C6H14O
CH3(CH2)5OH
100 mL
0.8136
1-hexanol, hexyl alcohol, hexanol, n-hexanol, n-hexyl alcohol, 1-hexyl alcohol, 1-hydroxyhexane, amylcarbinol, caproyl alcohol, pentylcarbinol
ZSIAUFGUXNUGDI-UHFFFAOYSA-N
hexan-1-ol
8103
CHEBI:87393
98%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Warning

EINECSNumber : 203-852-3

SDS
Documents

Documents

RUO – Research Use Only