missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hexylbenzene 98.0+%, TCI America™
$146.14 - $1,096.13
Chemical Identifiers
CAS | 1077-16-3 |
---|---|
Molecular Formula | C12H18 |
Molecular Weight (g/mol) | 162.28 |
MDL Number | MFCD00009526 |
InChI Key | LTEQMZWBSYACLV-UHFFFAOYSA-N |
Synonym | 1-phenylhexane, n-hexylbenzene, benzene, hexyl, hexane, 1-phenyl, unii-8e3d1a2bzp, 8e3d1a2bzp, phenylhexane, hexyl-benzen, 1-hexylbenzene, 4-hexylbenzene |
PubChem CID | 14109 |
IUPAC Name | hexylbenzene |
SMILES | CCCCCCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
H013625ML
|
TCI America
H013625ML |
25 mL |
N/A
|
|
|||||
H0136250ML
|
TCI America
H0136250ML |
250 mL |
N/A
|
|
|||||
Chemical Identifiers
1077-16-3 | |
162.28 | |
LTEQMZWBSYACLV-UHFFFAOYSA-N | |
14109 | |
CCCCCCC1=CC=CC=C1 |
C12H18 | |
MFCD00009526 | |
1-phenylhexane, n-hexylbenzene, benzene, hexyl, hexane, 1-phenyl, unii-8e3d1a2bzp, 8e3d1a2bzp, phenylhexane, hexyl-benzen, 1-hexylbenzene, 4-hexylbenzene | |
hexylbenzene |
Specifications
1077-16-3 | |
Colorless | |
C12H18 | |
25 mL | |
1-phenylhexane, n-hexylbenzene, benzene, hexyl, hexane, 1-phenyl, unii-8e3d1a2bzp, 8e3d1a2bzp, phenylhexane, hexyl-benzen, 1-hexylbenzene, 4-hexylbenzene | |
CCCCCCC1=CC=CC=C1 | |
162.28 | |
162.28 | |
Liquid |
-61°C | |
226°C | |
MFCD00009526 | |
3082 | |
LTEQMZWBSYACLV-UHFFFAOYSA-N | |
hexylbenzene | |
14109 | |
≥98.0% (GC) | |
Hexylbenzene |
Safety and Handling
EINECSNumber : (3)-0021
TSCA : Yes