missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexylcyclohexane 98.0+%, TCI America™

$264.65 - $519.43

Chemical Identifiers

CAS 4292-75-5
Molecular Formula C12H24
Molecular Weight (g/mol) 168.32
MDL Number MFCD00019403
InChI Key QHWAQXOSHHKCFK-UHFFFAOYSA-N
Synonym 1-Cyclohexylhexane
PubChem CID 20283
IUPAC Name hexylcyclohexane
SMILES CCCCCCC1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H013710ML
View Documents
TCI America
H013710ML
10 mL
Each for $264.65
 
H013725ML
View Documents
TCI America
H013725ML
25 mL
Each for $519.43
 
Description

Description

Specifications

Chemical Identifiers

4292-75-5
168.32
QHWAQXOSHHKCFK-UHFFFAOYSA-N
20283
CCCCCCC1CCCCC1
C12H24
MFCD00019403
1-Cyclohexylhexane
hexylcyclohexane

Specifications

4292-75-5
221°C
MFCD00019403
1-Cyclohexylhexane
CCCCCCC1CCCCC1
168.32
168.32
Liquid
Colorless
C12H24
10 mL
QHWAQXOSHHKCFK-UHFFFAOYSA-N
hexylcyclohexane
20283
≥98.0% (GC)
Hexylcyclohexane
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'