missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexylene Glycol, Reagent, 98%, Spectrum™ Chemical

HE188, 107-41-5, C6H14O2

Supplier:  Spectrum Chemical Mfg Cor HE1884LT

 View more versions of this product

Catalog No. 18603130


Explore available promotions

Description

Description

Spectrum™ Chemical Hexylene Glycol, Reagent
Specifications

Specifications

107-41-5
Amber Glass Bottle
4 L
CC(O)CC(C)(C)O
118.18
Reagent
100%
C6H14O2
SVTBMSDMJJWYQN-UHFFFAOYNA-N
2-methylpentane-2,4-diol
98%

Chemical Identifiers

107-41-5
118.18
2-methylpentane-2,4-diol
C6H14O2
SVTBMSDMJJWYQN-UHFFFAOYNA-N
CC(O)CC(C)(C)O
SDS
Documents

Documents

Promotions

Promotions