Learn More
Hexylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals
$126.27 - $126.27
Chemical Identifiers
| CAS | 3761-92-0 |
|---|---|
| Molecular Formula | C6H13BrMg |
| Molecular Weight (g/mol) | 189.38 |
| MDL Number | MFCD00000046 |
| InChI Key | SFXUWNMDTWQSHE-UHFFFAOYSA-M |
| Synonym | hexylmagnesium bromide, magnesium, bromohexyl, n-hexylmagnesium bromide, n-hexmgbr, hexyl magnesium bromide, n-hexyl magnesium bromide, hexylmagnesium bromide solution, sfxuwnmdtwqshe-uhfffaoysa-m, hexylmagnesium bromide solution, 2.0 m in diethyl ether, grignard reagent |
| PubChem CID | 4184939 |
| IUPAC Name | magnesium;hexane;bromide |
| SMILES | CCCCCC[Mg]Br |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC427761000
|
Thermo Scientific Chemicals
427761000 |
100 mL | AcroSeal™ Glass bottle |
Each for $126.27
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3761-92-0 | |
| 189.38 | |
| SFXUWNMDTWQSHE-UHFFFAOYSA-M | |
| 4184939 | |
| CCCCCC[Mg]Br |
| C6H13BrMg | |
| MFCD00000046 | |
| hexylmagnesium bromide, magnesium, bromohexyl, n-hexylmagnesium bromide, n-hexmgbr, hexyl magnesium bromide, n-hexyl magnesium bromide, hexylmagnesium bromide solution, sfxuwnmdtwqshe-uhfffaoysa-m, hexylmagnesium bromide solution, 2.0 m in diethyl ether, grignard reagent | |
| magnesium;hexane;bromide |
Specifications
| 3761-92-0,109-99-9 | |
| 0.9800g/mL | |
| C6H13BrMg | |
| MFCD00000046 | |
| 0.98 | |
| SFXUWNMDTWQSHE-UHFFFAOYSA-M | |
| magnesium;hexane;bromide | |
| 4184939 | |
| Liquid |
| Brown | |
| AcroSeal™ Glass bottle | |
| CH3(CH2)5MgBr | |
| 100 mL | |
| hexylmagnesium bromide, magnesium, bromohexyl, n-hexylmagnesium bromide, n-hexmgbr, hexyl magnesium bromide, n-hexyl magnesium bromide, hexylmagnesium bromide solution, sfxuwnmdtwqshe-uhfffaoysa-m, hexylmagnesium bromide solution, 2.0 m in diethyl ether, grignard reagent | |
| CCCCCC[Mg]Br | |
| 189.38 | |
| 189.39 | |
| Hexylmagnesium bromide, 0.8M solution in THF |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
RUO – Research Use Only