missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hexyltrimethylammonium Bromide 98.0+%, TCI America™

$188.59 - $1461.94

Chemical Identifiers

CAS 2650-53-5
Molecular Formula C9H22BrN
Molecular Weight (g/mol) 224.186
MDL Number MFCD00051872
InChI Key JYVPKRHOTGQJSE-UHFFFAOYSA-M
Synonym hexyltrimethylammonium bromide, hexyltrimethylazanium bromide, acmc-209grw, hexyltrimethylammoniumbromide, n-hexyltrimethylammonium bromide, n,n,n,n-hexyltrimethylammonium bromide, 1-hexanaminium,n,n-trimethyl-, bromide, 1-hexyl trimethylammonium bromide, 1-hexanaminium,n,n,n-trimethyl-, bromide 1:1
PubChem CID 10059492
IUPAC Name hexyl(trimethyl)azanium;bromide
SMILES CCCCCC[N+](C)(C)C.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H053425G
View Documents
TCI America
H053425G
25 g
Each for $188.59
 
H0534500G
View Documents
TCI America
H0534500G
500 g
Each for $1,461.94
 
Description

Description

Specifications

Chemical Identifiers

2650-53-5
224.186
JYVPKRHOTGQJSE-UHFFFAOYSA-M
10059492
CCCCCC[N+](C)(C)C.[Br-]
C9H22BrN
MFCD00051872
hexyltrimethylammonium bromide, hexyltrimethylazanium bromide, acmc-209grw, hexyltrimethylammoniumbromide, n-hexyltrimethylammonium bromide, n,n,n,n-hexyltrimethylammonium bromide, 1-hexanaminium,n,n-trimethyl-, bromide, 1-hexyl trimethylammonium bromide, 1-hexanaminium,n,n,n-trimethyl-, bromide 1:1
hexyl(trimethyl)azanium;bromide

Specifications

2650-53-5
White
MFCD00051872
hexyltrimethylammonium bromide, hexyltrimethylazanium bromide, acmc-209grw, hexyltrimethylammoniumbromide, n-hexyltrimethylammonium bromide, n,n,n,n-hexyltrimethylammonium bromide, 1-hexanaminium,n,n-trimethyl-, bromide, 1-hexyl trimethylammonium bromide, 1-hexanaminium,n,n,n-trimethyl-, bromide 1:1
CCCCCC[N+](C)(C)C.[Br-]
224.186
224.19
Crystalline Powder
185°C
C9H22BrN
25 g
JYVPKRHOTGQJSE-UHFFFAOYSA-M
hexyl(trimethyl)azanium;bromide
10059492
≥98.0% (HPLC,T)
Hexyltrimethylammonium Bromide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'