Mopec HF100 Organ Rinse Dissecting Station with Large Sink
Allow for multiple dissecting boards to be placed next to each other directly in front of the subdivided long rinse sink. Mopec™ HF100 Organ Rinse Dissecting Station with Large Sink includes eight removable dividers to allow the separation of organs during the rinsing process.
Supplier:  Mopec HF100
Catalog No. 22444114
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • All stainless steel construction
  • Eight foot long dissecting areas
  • Removable perforated dividers provide up to eight sectioned compartments
  • 72 inch long sink with overflow drain
Warranty
1 Year Warranty
Specifications
Specifications
Dissection Station
97 in.
37 in.
86.36 cm
182.88 cm
Stainless Steel
246.38 cm
34 in.
72 in.
93.98 cm