Applied Biosystems™ Hi-Di™ Formamide DFS Item
Highly deionized (Hi-Di) formamide
Supplier:  Applied Biosystems™ 4311320
 View more versions of this product
Catalog No. 4311320
Explore available promotions
Description
Description
Highly deionized (Hi-Di) formamide is used to resuspend samples before electrokinetic injection in capillary electrophoresis systems.
Specifications
Specifications
Store at -15°C to -25°C. Avoid multiple freeze-thaw cycles.
High-Di™
25 mL
Dry Ice
Sequencing
Formamide
Formamide
Liquid
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.