Applied Biosystems™ Hi-Di™ Formamide
Highly deionized (Hi-Di) formamide
Supplier: Applied Biosystems™ 4440753
Description
- Highly deionized (Hi-Di) formamide is used to resuspend samples before electrokinetic injection in capillary electrophoresis systems.
- The smaller configuration (5mL) minimizes the number of freeze/thaw cycles compared to the larger 25mL bottle
Order Info
Each
Specifications
High-Di™ | |
Formamide, 4x 5mL, Liquid, Formamide Reagent Type | |
4x 5 mL |
Store at -15 to -25°C. | |
Formamide |
For Research Use Only. Not for use in diagnostic procedures.